VWR® Reclosable Bags with Writing Area
Supplier: VWR, part of Avantor
VWR®
3970 3960 4000A 3945A 3950 3940A 3970A 3960A 3985A 3990A 3945
89073-138CS 95.07 USD
89073-138 89073-146 89073-148 89071-872 89071-874 89073-154 89071-876 89073-144 89071-878 89073-150 89073-140
VWR® Reclosable Bags with Writing Area
Bags Storage Bags
Reclosable bags are full gauge and offer exceptional clarity and quality.
  • Reclosable design
  • White writing area is compatible with markers or pens
  • Reinforced zipper locks in a smooth motion
Red line bags have a red line along the top of the bag. Made of 100% virgin Low Density Polyethylene (LDPE) resin.
Certificaciones: Meet FDA/USDA specifications.