VWR® Meker Type High-Temperature Burners
Supplier: VWR, part of Avantor
VWR®
89038-546 89038-544
89038-546CS 2047.68 USD
89038-546 89038-544
VWR® Meker Type High-Temperature Burners
Heaters Gas Burners
Meker-type high-temperature burners are for use where steady high temperatures are required.
  • Backflashes eliminated via safety cap feature
  • Heavy die-cast base
  • Uniform, consistent temperature
The 8.6 cm (33/8") Meker grid top forms short, uniform blue cones of consistent temperature. Safety cap design eliminates backflashes. Heavy die-cast base unit and venturi tube are made from nickel-plated zinc alloy. Overall height: 21.6 cm (81/2"). Grid diameter: 3.8 cm (11/2").