VWR® Adjustable Tirrill Burners
Supplier: VWR, part of Avantor
VWR®
89038-538 89038-536 89038-540
89038-538CS 673.79 USD
89038-538 89038-536 89038-540
VWR® Adjustable Tirrill Burners
Heaters Gas Burners
Burners are ideal for separate control of flame temperature and height.
  • Fine-tuned control of gas intake
  • Total height of 8.9 cm
Burner provides clear flame from 1.3–30.5 cm (1/2–12"). Gas intake is controlled by a fine-pitch brass needle valve. Overall height: 8.9 cm (31/2"). Optional wing top produces a broad, flat flame for working glass and general lab use.
Información para pedidos: Shipped complete. Wing top adaptor is included with all burners except 89038-536, and must be ordered separately for this unit.