VWR® Bunsen Burners
Supplier: VWR, part of Avantor
VWR®
89038-532 89038-530
89038-532CS 665.62 USD
89038-532 89038-530
VWR® Bunsen Burners
Heaters Gas Burners
Designed with a flame stabilizer to provide a steady flame for general laboratory needs.
  • Stable, heavy base
  • Steady flame
Stable, heavyweight die-cast base of nickel-plated zinc-alloy with aluminum mixing tube, 8mm (5/16") serrated gas inlet, and air regulator. The optional wing top produces a broad flat flame for working glass and general lab use.
Información para pedidos: Shipped complete. The wing top must be ordered separately.