VWR® PTFE Vials
Supplier: VWR, part of Avantor
VWR®
014.125 014.150
89026-430EA 323.73 USD
89026-430 89026-432
VWR® PTFE Vials
Vials
PTFE vials are ideal for storing aggressive or valuable materials.
Suitable for small-scale reactions at low pressure. Vials feature an inner taper for easy removal of contents and thick wall construction for durability.