VWR® Stirring Rods
Supplier: VWR, part of Avantor
VWR®
003.250 003.100 003.400 003.200 003.300
89026-286PK 232.24 USD
89026-286 89026-284 89026-282 89026-280 89026-290
VWR® Stirring Rods
Stirrers Stirring Rods
Available with a steel core or in solid PTFE, these rods are chemically resistant, inert, and non-abrasive.
  • Constructed from solid PTFE or with a steel core
  • Chemically resistant
  • Inert
  • Non-abrasive
Steel core rods have a rounded end, can be bent into a permanent shape, and are ideal when extra rigidity is required. Solid PTFE rods have a tapered end. Usable to 280°C (536°F).