VWR® HDPE Multipurpose Containers
Supplier: VWR, part of Avantor
VWR®
0417-1100 0404-1100 0485-1100 0495-1100 0464-1100 0432-1100 0490-1100 0408-1100
89009-668CS 224.79 USD
89009-668 89038-282 89009-666 89009-664 89009-674 89009-662 89009-672 89009-670
VWR® HDPE Multipurpose Containers
Sample Containers General Purpose Containers
Translucent containers for the storage of pathology, industrial, and environmental samples.
  • HDPE construction
  • Chemical-resistant
  • Translucent siding
Containers are made of high-quality plastic to resist breakage and come in a wide range of sizes. The matching cap snaps on tight for a secure fit. The 4800 mL container is equipped with a handle for portability.