KIMAX® Burets, Class B, Straight Bore Stopcock, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
17026F-25 17026F-50 17026F-100 17026-50 17026F-10
89001-608CS 217.9 USD
89001-608 89001-606 89001-614 89001-612 89001-610
KIMAX® Burets, Class B, Straight Bore Stopcock, Kimble Chase
Burets
Burets feature a 2mm stopcock plug and a KIM-KAP® dust cap
Scale is printed in durable black ceramic enamel. Designed from ASTM Specification E287, Class B requirements.