VWR® PureStep Adhesive Mats
Supplier: VWR, part of Avantor
VWR®
PS 2436 38 W PF203801W00 PS 2645 38 G PS 2430 34 W PS 1836 64 W PS 2430 64 W PS 2430 64 G PF383801W00 PS 3660 34 W PS366034W-HT PF284701W00 PS183638W-HT PS184538B-HT PS 3645 38 B PS 2645 38 B PS 2430 64 B PS 1845 38 G PS 1836 38 W PS 1845 38 B PS 3636 38 W PS 1845 38 W PS 3645 64 G PS 2436 64 B PS 3636 64 W PS 3645 38 W PS 2436 64 W PS 3636 38 B PS 2436 38 B PS 2645 64 W PS 3636 38 G PS 2645 64 G PF386201W00 PS 1836 38 B PS366064B PS 2430 34 G PS 2645 38 W PS 3636 64 G PS2430320C-LT PS 1845 64 W PS 3645 38 G PS243638B-HT PS 2645 64 B PS 2436 64 G PS364564W-HT PS 1845 64 B PS 3645 64 W PS366034B-HT PS 2430 34 B 254564W-HT PF204701W00 PS 1836 38 G PS 1836 64 G PS 3636 64 B PS264538BHT PS264564BHT PS 3660 34 B PS 1836 64 B PF263801W00 PF384701W00 PS 1845 64 G PS 3660 34 G PS364538BHT PS 2436 38 G PS 2430 34 C PS 3645 64 B PS264564W-HT
87004-018CS 339.01 USD
87004-018 87004-016 87004-014 87004-058 87004-012 87004-056 87004-298 87004-054 87004-296 87004-052 89126-768 87004-050 87004-092 87004-090 89126-772 82031-466 89126-770 87004-008 87004-028 87004-026 87004-068 87004-024 87004-300 87004-066 87004-022 87004-020 87004-064 87004-062 87004-060 89131-500 82029-678 82029-680 89428-992 87004-038 87004-036 87004-078 87004-034 87004-076 87004-032 87004-030 87004-074 87004-072 87004-070 87004-006 82028-392 87004-004 87004-048 87004-046 87004-002 87004-044 87004-000 87004-088 82028-394 87004-086 87004-042 89066-232 87004-084 87004-040 87004-082 87004-080 82031-676 89126-764 89126-766 89126-760 89126-762 89089-166
VWR® PureStep Adhesive Mats
Floor Mats Tacky Mats
Multilayered contamination control mats remove and contain dirt and dust from foot traffic and equipment wheels.
  • 0.3 mil thick acrylic-based, pressure-sensitive adhesive coating
  • Antimicrobial agent protects against growth of bacteria, mold, and fungus
Mats consist of 30 or 60 sheets of 1.5 mil low-density polyethylene film. Each sheet has a 0.3 mil thick acrylic-based, pressure-sensitive adhesive coating. Numbered tabs ensure one-at-a-time sheet removal. Mats can easily be removed during routine floor maintenance. Some sizes are available with high-tack adhesives. Mat frames fit mats of sizes specified. Frames provide added stability with nonskid backing and allow use on surfaces that are not ideal for tacky mats.
Información para pedidos: Mats and frames are sold separately. For adhesive mats pre-applied to single or double frames, see the VWR® Intro Mats with Stabilizing Frame series.