VWR® Signature™ Single-Channel Pipettors, Fixed Volume
Supplier: VWR, part of Avantor
VWR® Signature™
P3820-50VWR P3820-5VWR P3820-1000VW P3820-25VWR P3820-500VWR P3820-10VWR P3820-200VWR P3820-100VWR P3820-20VWR P3820-250VWR
83009-770EA 270.61 USD
83009-770 83009-760 83009-772 83009-762 83009-774 83009-764 83009-776 83009-766 83009-768 83009-758
VWR® Signature™ Single-Channel Pipettors, Fixed Volume
Pipets Micropipets
Ideal for applications where specific volumes are used repeatedly, these slender, contoured pipettor bodies fit comfortably in either hand, as well as in large or small hands.
  • Ten fixed volumes available
  • Narrow shaft for universal tip fit
  • Removable, adjustable tip ejector
  • Fully autoclavable
  • VWR® two-year limited warranty
  • Pipettor stands hold six pipettors
In addition, maintenance-free seals and highly polished pistons provide exceptional accuracy and precision, as well as allow the pipettors to be autoclaved repeatedly without recalibration
The corrosion-resistant, stainless steel tip ejector is removable for pipetting into narrow tubes and it can be adjusted to accommodate various types of tips. The slender, tapered shaft provides universal tip compatibility and is highly chemical-resistant.