VITLAB® Laboratory Bottles, LDPE, Wide Mouth, with Screw Caps, PP, BrandTech
Supplier: BrandTech
V93989 V94189
83009-062PK 175.47 USD
83009-062 83009-063
VITLAB® Laboratory Bottles, LDPE, Wide Mouth, with Screw Caps, PP, BrandTech
Bottles Roller Bottles
Sqaure-shouldered general-use low-density polyethylene (LDPE) laboratory bottles come with polypropylene (PP) screw caps.
  • Excellent chemical resistance to aliphatic alcohols, aldehydes, alkalis, and acids
  • Wide mouth design allows easy access to contents
For replacement caps, see 83009-032 series.