VITLAB® Square Laboratory Bottles, HDPE, Wide Mouth, with PP Screw Caps, BrandTech
Supplier: BrandTech
V92789 V92889 V92489 V92689
83009-055PK 232.92 USD
83009-055 83009-056 83009-057 83009-058
VITLAB® Square Laboratory Bottles, HDPE, Wide Mouth, with PP Screw Caps, BrandTech
Bottles Roller Bottles
General-use, high-density polyethylene laboratory bottles come with polypropylene screw caps.
  • Feature a space-saving square shape
  • Wide mouth design provides excellent access to bottle contents
Bottles provide excellent chemical resistance to aliphatic and aromatic hydrocarbons, alcohols, aldehydes, alkalis, and acids.