Polypropylene Screw Caps, BrandTech
Supplier: BrandTech
V83313 V83310 V83312
83009-032PK 54.97 USD
83009-032 83009-034 83009-035
Polypropylene Screw Caps, BrandTech
Closures Bottle Closures
Translucent screw caps are for use with BrandTech polypropylene and low-density polyethylene bottles.
  • Autoclavable translucent polypropylene
  • Fits VITLAB and many other bottles with standard thread
Envase: Bulk packed.