VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech
Supplier: BrandTech
V68894 V69294 V69194 V68594
83009-022PK 72.78 USD
83009-022 83009-023 83009-016 83009-019
VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech
Vials
General-purpose sample vials for a variety of lab applications.
  • Polypropylene
  • LDPE push-on 'snap' caps
  • Vials are autoclavable, caps are not
Translucent polypropylene vials include low-density polyethylene push-on caps. Vials and caps are suitable for microwave use.