VITLAB® Forceps, POM, BrandTech
Supplier: BrandTech
V68099 V68399 V68199 V68299
83009-012PK 28.7 USD
83009-012 83009-013 83009-014 83009-015
VITLAB® Forceps, POM, BrandTech
Forceps
Polyoxymethylene (POM) forceps with rounded ends.
  • Rounded ends
  • Opaque orange-yellow
POM is suitable for autoclaving but not suitable for microwaves. Frequent autoclaving may reduce mechanical stability. Resistant to aliphatic alcohols, ether, aldehydes, aliphatic hydrocarbons, aromatic hydrocarbons, halogenated hydrocarbons, ketones, and alkalis.
Operating temperature: –40 to 130°C (–40 to 266°F)
Precaución: Not recommended for use with ester, acids, and oxidizing agents.