VITLAB® Forceps, PMP, BrandTech
Supplier: BrandTech
V67895 V67995
83009-010PK 64.21 USD
83009-010 83009-011
VITLAB® Forceps, PMP, BrandTech
Forceps
Polymethylpentene (PMP) forceps with pointed ends.
  • Autoclavable at 121 °C for 20 minutes
  • Translucent
PMP is suitable for autoclaving and microwaves. Resistant to aliphatic alcohols, aldehydes, ester, aliphatic hydrocarbons, ketones, alkalis, and most acids (except oxidizing acids). Not recommended for use with ether, aromatic hydrocarbons, halogenated hydrocarbons, and oxidizing agents. Color: transparent white.
Operating temperature: 0 to 150°C (32 to 302°F)