VITLAB® Griffin Beakers, PMP, BrandTech
Supplier: BrandTech
V61803 V61603 V60803 V61503 V60903 V61103 V60703 V61203 V60503 V61703 V61403 V61003 V60603
83008-808PK 127.27 USD
83008-808 83008-780 83008-784 83008-792 83008-820 83008-832 83008-788 83008-796 83008-824 83008-800 83008-828 83008-804 83008-816
VITLAB® Griffin Beakers, PMP, BrandTech
Beakers
High-quality beakers made of plastic polymethylpentene with moderately good chemical resistance.
  • Glass-clear polymethyl pentene (PMP)
  • Red screened graduations for high contrast with many liquids
Beakers feature molded graduations with red-screened overprint for legibility.
Certificaciones: All beakers meet the requirements of ISO 7056.