VITLAB® Griffin Beakers, Polypropylene, BrandTech
Supplier: BrandTech
V607081 V609081 V608081 V605081 V617081 V616081 V615081 V614081 V606081 V612081 V610081 V611081 V618081
83008-790PK 57.99 USD
83008-790 83008-794 83008-782 83008-798 83008-810 83008-830 83008-786 83008-802 83008-822 83008-834 83008-806 83008-818 83008-826
VITLAB® Griffin Beakers, Polypropylene, BrandTech
Beakers
High quality beakers feature molded graduations (except 83008-814) with blue-screened overprint for legibility.
  • Highly chemically resistant polypropylene
  • High translucency
  • Blue screened graduations provide high contrast with many liquids
Beakers are translucent for compatibility and offer good chemical and break resistance.
Certificaciones: All beakers meet the requirements of ISO 7056.