VITLAB® Graduated Pitchers, Polypropylene, BrandTech
Supplier: BrandTech
V482941 V442081 V440941 V445941 V481941 V447941 V484941 V441941 V442941 V446081 V447081 V483941 V441081 V445081 V440081 V443081 V444941 V443941 V444081 V446941
83008-772PK 130.99 USD
83008-772 83008-762 83008-740 83008-770 83008-760 83008-776 83008-754 83008-766 83008-774 83008-730 83008-752 83008-742 83008-764 83008-736 83008-758 83008-748 83008-734 83008-778 83008-746 83008-768
VITLAB® Graduated Pitchers, Polypropylene, BrandTech
Beakers
Made of durable, translucent polypropylene.
  • Pitchers with molded graduations areavailable with or without overprint
  • Nesting pitchers have screened overprint
  • Blue printing provides high contrast with many liquids
Standard graduated pitchers have molded graduations with or without a blue overprint for high legibility. Nesting pitchers conserve bench and storage space and have blue screened graduations for easy, accurate measurements.