VITLAB® Graduated Pitchers, SAN, BrandTech
Supplier: BrandTech
V44291 V44491 V44391 V44191 V44091
83008-750PK 238.67 USD
83008-750 83008-732 83008-744 83008-756 83008-738
VITLAB® Graduated Pitchers, SAN, BrandTech
Beakers
Clear SAN (styrene acrylonitrile copolymer) pitchers with molded graduations are a lightweight, safer alternative to glass in many applications.
  • Glass-clear pitchers have contoured handles for easy lifting
  • Durable molded graduations for long measuring life
  • Pouring lip reduces drips
  • SAN has a maximum temperature for use of 70 ˚C
Enhanced graduations are provided at major intervals for easy volume estimations.