VITLAB® Griffin Beakers, ETFE, BrandTech
Supplier: BrandTech
V111004 V110304 V110804 V110604 V110704 V110904 V110204
83008-640PK 134.16 USD
83008-640 83008-644 83008-620 83008-632 83008-638 83008-624 83008-636
VITLAB® Griffin Beakers, ETFE, BrandTech
Beakers
Made of ETFE (ethylene tetrafluoroethylene), a fluoropolymer.
  • High chemical and thermal resistance
  • Graduated
Beakers are translucent with black graduations for easy volume estimation.
Certificaciones: Beakers meet the requirements of ISO 7056.