VITLAB® Low Form Griffin Beakers, PFA, BrandTech
Supplier: BrandTech
V110605 V110905 V110305 V111005 V110405 V110205
83008-630PK 142.08 USD
83008-630 83008-622 83008-634 83008-642 83008-626 83008-646
VITLAB® Low Form Griffin Beakers, PFA, BrandTech
Beakers
Beakers are made of PFA (perfluoroalkoxy copolymer), a fluoropolymer with excellent chemical resistance
  • Containers provide excellent stability for standards and are manufactured without metal catalysts
  • Highly translucent for verification of contents
  • Thermally stable from –200 to 260°C (–328 to 500°F)
  • Molded graduations are easy-to-read and resistant to wear from repeated use
  • Autoclavable
Surfaces are much less porous than PTFE and are highly cleanable to reduce sample contamination by “memory effect” from prior samples. Ideal for use in all aspects of ICP-MS and other trace analysis work.
Certificaciones: Beakers conform to ISO 7056.