VITLAB® Volumetric Flasks with Screw Caps, PFA, BrandTech
Supplier: BrandTech
V107597 V107497 V107197 V107097 V107297 V107397
83008-574PK 1080.7 USD
83008-574 83008-572 83008-568 83008-566 89093-094 83008-570
VITLAB® Volumetric Flasks with Screw Caps, PFA, BrandTech
Flasks Volumetric Flasks
Flasks feature a durable, easy-to-read, individually calibrated ring marking.
  • PFA for extreme chemical resistance
  • Screw caps help maintain sample integrity
Highly translucent, hydrophobic surfaces ensure easy meniscus verification. Made of PFA (perfluoroalkoxy copolymer). Highly heat- and chemical-resistant, autoclavable, and easy-to-clean. Can withstand temperatures from –200 to 260 °C (–328 to 500 °F). PFA screw caps help protect against outside contamination.
Certificaciones: Flasks meet Class A tolerances per DIN EN ISO 1042.