VITLAB® Reagent Bottles, with PP Screw Caps, BrandTech
Supplier: BrandTech
V100689 V100889 V100989 V100789 V100489 V100389
83008-530PK 66.43 USD
83008-530 83008-534 83008-510 83008-522 83008-514 83008-526
VITLAB® Reagent Bottles, with PP Screw Caps, BrandTech
Bottles Plastic Bottles
Slope-shouldered polypropylene bottles feature polypropylene screw caps.
  • Bottles and caps are autoclavable
They provide excellent chemical resistance to aliphatic alcohols, aldehydes, aliphatic hydrocarbons, alkalis, and acids.
Información para pedidos: For replacement caps, see 83009-032 series.
Información de suministro: 10 liter size includes two handles, five liter size includes a handle to facilitate movement.