Puritan® Foam-Over-Cotton Tipped Swab, Wood Handle, Puritan Medical Products
Supplier: Puritan Medical Products
Puritan®
1806-WCF 1806-WCSFB 1806-WCFB 1806-WCSFB 1806-WCFB 1806-WCF
82030-594CS 543.62 USD
CA82030-594 82030-594 CA82030-592 82030-592 CA82030-590 82030-590
Puritan® Foam-Over-Cotton Tipped Swab, Wood Handle, Puritan Medical Products
Swabs Swabs for Sampling
This absorbent foam tipped applicator is produced with reticulated 100 ppi medical grade foam over high quality USP grade cotton.
It is bonded securely to a rigid white birch shaft. For use in critical environments and as cleaning applicators for particle removal.