VWR® Crucible Tongs
Supplier: VWR, part of Avantor
VWR®
11648-104 11648-101
82027-362CS 866.29 USD
82027-362 82027-368
VWR® Crucible Tongs
Tongs
Crucible tongs are designed for use in handling a wide variety of laboratory apparatus.
  • Nickel-plated stainless steel fabrication
The 24.1cm model is a general-purpose tong with serrated tips. The 45.7cm model features nickel-plated steel construction and an extra-long design.