VWR® Small Latex Bulbs
Supplier: VWR, part of Avantor
VWR®
HS20622BV HS20622AV
82024-550CS 7835.65 USD
82024-550 82024-554
VWR® Small Latex Bulbs
Pipets Pipet Filler Bulbs
Small-volume, natural rubber latex bulbs are designed for use with Pasteur pipets, small pipets, and medicine droppers.
  • Small-volume design
  • Natural rubber latex construction
  • White finish
Supplied with white finish.