VWR® Calibrated Electronic Thermometers with Waterproof Sensor
Supplier: VWR, part of Avantor
VWR®
60900-0211 60900-0811 60900-2711 60900-2411
98000-208EA 51.94 USD
98000-208 98000-206 82021-172 82020-064
VWR® Calibrated Electronic Thermometers with Waterproof Sensor
Thermometers Digital Thermometers
Digital thermometers that effectively measure the temperature of wet or dry environments including freezers, refrigerators, water baths, aquariums, incubators and more.
  • Features available include minimum/maximum temperature readings, dual/triple display, and alarms
  • All models include accredited calibration documentation that meets the requirements of ISO 17025
  • Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish)
  • Individually serialized, CE marked
Tested and calibrated in an exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.