VWR® PCR Plates
Supplier: VWR International
3971-520-300
82006-650CS 775.32 USD
82006-650
VWR® PCR Plates
Microplates PCR Plates
VWR PCR plates are designed to fit most popular thermocyclers.
  • Polypropylene construction
  • Autoclavable
  • Alphanumeric grid
Each well holds 0.1 mL in a low profile format that fits more precisely with newer thermocycling engines
lncludes printed alpha-numeric grid. Thin wall low-profile design fits most popular Thermo-cyclers.
Extra thin walls ensure proper heat distribution. Certified to be free of Endotoxin, Oleamide, DNA and Nuclease (RNase/DNase).
Certificaciones: ISO 9001, CE IVD
Información para pedidos: Packed 10 tubes per box. 10 boxes per case.