BRAND 96-Well and 384-Well Deep Well Plates, BrandTech
Supplier: BrandTech
DW2250 701352 701350 701346 DW2200 701354 701355 701340
76183-226PK 837.34 USD
76183-226 76183-224 80087-070 76181-496 89093-076 80087-068 80087-066 80087-064
BRAND 96-Well and 384-Well Deep Well Plates, BrandTech
Microplates Deep Well Microplates
Deep well plates are excellent for high throughput screening (HTS), assays requiring a mother plate, cell and tissue culture, and sample storage
  • Suitable for storage, reagent reservoirs or use as mother plates
  • Work with most robotic/lquid handling systems
  • Polypropylene modes suitable for –80 °C (–112 °F) cryostorage
Polypropylene plates are manufactured from high-quality virgin polypropylene, are autoclavable to 121 °C (250 °F), and are resistant to a wide variety of chemicals, including phenols, chloroform, and DMSO. Plates are compatible with most robotic samplers and automated liquid handling systems. Polypropylene plates withstand temperatures as low as –80 °C (–112 °F) and are suitable for cryo-storage. Polystyrene plate withstands temperatures as low as –20 °C (–4 °F), making it suitable for cold room applications. Wells are alphanumerically labeled and have notched corners to facilitate orientation in automated systems.
Información para pedidos: Aluminum sealing film 80087-180 is RNase- and DNase-free and ideal for cryo-storage.