BRAND® Tip Boxes for BRAND® Bulk Tips and Tip Refills, BrandTech
Supplier: BrandTech
Brand®
732990 732994 732996 732992
76534-690EA 8.48 USD
76534-690 76534-688 76534-686 76534-684
BRAND® Tip Boxes for BRAND® Bulk Tips and Tip Refills, BrandTech
Boxes Storage Boxes
Empty polypropylene tip boxes with carrier plates to fit BRAND® pipette tips.
  • Refillable
  • Autoclavable
  • Color-coded for easy identification
BRAND® Tip Boxes are designed to hold BRAND® bulk tips and Tip Refills. Carrier plate is included for bulk tips. Gray carrier plate is designed to fit nano-cap™, ultra-micro, and 50 µl tips, in both unfiltered and filtered variations. Yellow carrier plate is designed to fit BRAND 200 µl tips. Green carrier plate is for use with 300 µl tips. The box with blue carrier plate has extra height to fit 1000 µl tips, but will accept carrier plates from other tip sizes. Box and carrier plate are fully autoclavable at 121 °C for 20 minutes for sterilization.
Información para pedidos: Box/carrier plate is sold individually.
Envase: Recyclable packaging.