VWR® Economy Polypropylene Specimen Containers
Supplier: VWR, part of Avantor
Q104 Q104NS Q105 Q101 Q101NS Q102
76532-300CS 128.8 USD
76532-300 76532-298 76532-304 76532-302 76532-296 76532-294
VWR® Economy Polypropylene Specimen Containers
Sample Containers General Purpose Containers
Sturdy, shatter-resistant containers are manufactured from transparent polypropylene to allow constant sample viewing.
  • Sturdy, shatter-resistant design
  • Transparent siding
  • Polypropylene construction
  • Available in nonsterile or sterile formats
  • Containers feature molded graduations in 10 mL and 0.5 oz. increments
  • Polyethylene snap-on lids are sold separately, except where noted
Lids create a secure seal, and offer space for date, name, doctor, and room number molded into the surface. Lids fit both 5 and 8 oz. containers.
Certificaciones: Manufactured under EN ISO13485:2016.