VWR® Blue Nitrile Examination Gloves
Supplier: VWR, part of Avantor
VWR®
14024340 14024338 14024341 14024339 14024337
76518-334CS 150.09 USD
76518-334 76518-336 76518-342 76518-340 76518-338
VWR® Blue Nitrile Examination Gloves
Gloves
Ambidextrous gloves are soft and comfortable for everyday use. They protect hands from a broad range of chemicals. Beaded cuff allows easy donning.
  • Finger textured
  • Ambidextrous
  • Non sterile
  • Soft and comfortable
  • Protects hands from broad range of chemicals and hazards
  • Powder-free
Ideal for use in industrial and laboratory applications.
Made of premium compounded nitrile for high manufacturing quality measures and product consistency. Gloves protect against biohazards and a broad range of chemicals. Composition is 100% latex-free to eliminate adverse reactions associated with natural rubber latex proteins. Finger textured gloves feature rolled, beaded cuffs.
Certificaciones: Meet or exceed ASTM D6319 on water leak and dimension. Manufactured under GMP, ISO9001, ISO13485 and MDSAP quality system.
Envase: 100 gloves per box, 10 boxes per case.