VWR® Signature™ Ergonomic High Performance Single-Channel Variable Volume Pipettors
Supplier: VWR, part of Avantor
VWR® Signature™
P3925-250VWR P3925-10000VWR 8901-WT P3925-2VWR P3925-100VWR P3925-200VWR 8952-WT P01555-VE05M P3925-SKB3-VWR P3925-5000VWR P01555-VE200 P01555-VE050 P3925-20VWR P3925-SKA3-VWR P3925-1000VWR P01555-VE020 8951-WT P01555-VE250 P3925-10VWR P01555-VE01K P3925-50VWR P3925-SKC4-VWR
10002-830EA 542.72 USD
10002-830 89079-962 89079-972 75788-458 89079-964 75788-456 89079-974 10002-834 89079-966 89079-976 10002-778 89079-968 89079-978 10002-782 89079-960 10002-780 89079-970 75788-460 76498-676 10002-836 76498-678 76498-674
VWR® Signature™ Ergonomic High Performance Single-Channel Variable Volume Pipettors
Pipets Micropipets
The new, ergonomic design of the pipettors allow for consistently comfortable pipetting even with extended use.
  • Superior accuracy and precision for critical applications
  • Reduced plunger forces
  • Fully autoclavable
  • Volume locking mechanism
An extended, curved finger rest allows the pipettor to balance on the index finger, and both plunger and volume-adjustment forces have been significantly reduced to eliminate the strain of repetitive pipetting.
To accommodate a wide variety of tips, the easy-grip ejector system provides adjustment of the ejector height. The stainless steel construction of the ejector prevents corrosion and bending. These lightweight, durable pipettors are fully autoclavable and are supplied with a tool for in-lab calibration and a three-year warranty.
Each starter kit is supplied with the pipette volumes listed in the table below and a 4-place linear stand.
Volume setting is continuously adjustable using either the micrometer wheel or dispenser button. A locking mechanism prevents accidental volume shift while the instrument is in use.