CONTACT SLIDE CHROM 2 20SLIDES PK20
Supplier: LIOFILCHEM, INC.
Contact Slide Chrom 2, ChromaticTM Coli Coliform / Plate Count Agar + TTC + Neutralizing, Flex Dip-slide with a chromogenic selective medium for detection of E. Coli/coliforms and non selective medium for total bacterial count, rtu device with two media solidified onto a plastic support, 20slides
Learn more About VWR
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com