CONTACT SLIDE 1 120SLIDES PK120
Supplier: LIOFILCHEM, INC.
Contact Slide 1, PCA+TTC+Neutralizing/VRBG Agar+Neutralizing Flex Dip-slide with selective medium for detection of Enterobacteriaceae, a ready-to-use device with 2 different media coated onto plastic support used for microbial monitoring of surfaces, liquids and non selective medium, Size: 120slides
Learn more About VWR
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com