VWR® Freezer and Ultra Low Freezer Verifictaion Thermometers
Supplier: VWR, part of Avantor
VWR®
61004-0011 61004-0311
76491-674CS 394.03 USD
76491-674 76491-670
VWR® Freezer and Ultra Low Freezer Verifictaion Thermometers
Thermometers Analog Thermometers
These mercury free thermometers are traceable to NIST and cover ranges from -90 to 25 °C .
  • Verify sample temperatures in freezers/ultra-low freezers
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • Thermometers contain organic liquid, or EnviroKleen™ certified Enviro-Safe® liquid; feature permanently fused markings and lead free glass
  • PFA coating safely contains glass and liquid in case of breakage
  • Manufactured and tested by our USA quality team; individually serialized
Insert the thermometer into the bottle and place near the samples for best results.
Certificaciones: Traceable to NIST.
Información de suministro: Supplied with multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy and traceability plus VELCRO® and magnetic attachments.