VWR® Blood Bank Thermometer
Supplier: VWR, part of Avantor
VWR®
60805-0011
76491-642CS 448.2 USD
76491-642
VWR® Blood Bank Thermometer
Thermometers Analog Thermometers
This thermometer has been designed to indicate critical temperature range.
  • Total immersion
  • Individual serial numbers
  • PTFE coating helps prevent liquid and glass contamination
The critical temperature range from 4 to 6 °C is marked with arrows.
Certificaciones: Traceable NIST. Envirokleen certified by ChemTel (an independent laboratory).