CytoCite™ BG100 Portable Fluorometer, AAT BIOQUEST INC
Supplier: AAT BIOQUEST INC
CBG100
76484-960EA 2421.32 USD
76484-960
CytoCite™ BG100 Portable Fluorometer, AAT BIOQUEST INC
Spectrophotometers and Spectrometers Fluorescence Spectrophotometers
The CytoCite™ Fluorometer BG100 is a cloud-integrated, palm-sized fluorometer designed for reading the ubiquitous FITC channel (Ex/Em = 480 nm/520 nm), with a sensitivity limit capable of detecting as little as 1 nM of fluorescein, making it highly optimal for running essential assays such as DNA and protein quantification.
  • High sensitivity
  • Assay sample volumes of as little as 1 µl
Información accesorios: Cytocite™ sample tubes are 500 µl thin-walled polypropylene tubes validated for use with Cytocite or Qubit Fluorometer.
Precaución: For research use only.