ESC Swabs, Liofilchem, Inc.
Supplier: Liofilchem
85603 85601 85606 85612 85604 85608 85614 85610 85605 85609 85602 85607 85621 85615
76479-578PK 181.41 USD
76479-578 76479-568 76479-572 76479-584 76479-562 76479-570 76479-582 76479-576 76479-566 76479-588 76479-574 76479-564 76479-586 76479-580
ESC Swabs, Liofilchem, Inc.
Swabs Swabs with Rinse Solutions
ESC (Easy Surface Checking) for verification and validation of cleaning procedures; cost effective and rapid method for microbial environmental monitoring.
  • Prefilled swab device
  • Rayon bud
  • Wide range of transport media and volumes
  • Leak-proof closure
  • Transport and storage at room temperature
ESC (Easy Surface Checking) swab range includes a new generation of sampling devices for microbiological examination of surfaces, especially recommended for food and pharmaceutical industries.
Certificaciones: Conform to international standards (ISO 18593, ISO 17604)