VWR® Critical Environment Swab, Large Flexible Foam Head, VWR International
Supplier: VWR, part of Avantor
VWR®
SU-113VFCTD
76478-842BX 40.51 USD
76478-842
VWR® Critical Environment Swab, Large Flexible Foam Head, VWR International
Swabs Swabs for Clinical Use
Swab features 100 ppi closed cell polyurethane foam with 100% virgin polypropylene handle.
  • Polyurethane foam swab head
  • Polypropylene handle
  • Large size head
Swab head is thermally bonded to the handle and augmented with adhesive.