SP Bel-Art Sterileware® Powder Funnels, Bel-Art Products, a part of SP
Supplier: Bel-Art
Sterileware®
F14660-1100 F14660-1065 F14660-1150 F14660-1080
76469-392CS 316.94 USD
76469-392 76469-398 76469-394 76469-396
SP Bel-Art Sterileware® Powder Funnels, Bel-Art Products, a part of SP
Funnels Powder Funnels
These sterile, wide stem funnels are ideal for transferring powders or granules using a sterile sampling tool into a sterile container.
  • 60° angle and large bore stem help minimize bridging of powder
  • FDA grade polypropylene; safe for food, drug and cosmetic use
  • Gamma irradiated sterile (SAL 10⁻⁶)
  • Individually bagged; ready for one time use and disposal
Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization.