VWR® A-Series Balances with Calibration Certificate
Supplier: VWR, part of Avantor
VWR®
VWR-520AC/CAL VWR-310AC/CAL VWR-363AC/CAL VWR-8102AC/CAL VWR-205AC/CAL VWR-603AC/CAL VWR-2102AC/CAL VWR-4502AC/CAL VWR-10102AC/CL VWR-105AC/CAL VWR-160AC/CAL VWR-6002AC/CAL VWR-1003AC/CAL VWR-220AC/CAL
76458-528EA 4588.8 USD
76458-528 76458-526 76329-734 76458-530 76458-520 76329-736 76329-740 76329-742 76329-962 76458-518 76458-522 76329-960 76329-738 76458-524
VWR® A-Series Balances with Calibration Certificate
Balances and Scales
The VWR® A-Series balances with calibration certificate are the answer for an organizations regulatory and compliance needs.
  • Complete calibration data reported
  • 2× RS 232, USB (A and B), and Wi-Fi communication interfaces
  • Five year warranty
  • Internal calibration
  • Easy-to-read, adjustable, backlit display
  • Tool-free assembly and diassembly of the 3-door draft shield on analytical models
The VWR® A-Series balances with calibration certificate include an ISO 17025:2017 certificate of calibration showing the full testing data using NIST-traceable weights. Featuring internal calibration, a multitude of communication options, and built in data protections, the VWR A-Series provide advanced features for any application. The large, back-lit LCD display allows for easy viewing of the weighing results. Data safety is provided via the internal memory allowing for recording up to 100,000 weighings. The analytical models feature a spacious 3-door draft shield that allows for tool-free assembly and disassembly of the weighing chamber. The VWR® A-Series balances with calibration certificate are the answer for organizations demanding precision and performance with full traceability.
Certificaciones: Includes an ISO/IEC 17025:2017 accredited Calibration and Testing Laboratory issues calibration certificate. CE marked.
Información para pedidos: Optional accessories such as printer (10799-132), density determination kit and granite platform (10799-162) are available to order separately. The 0.01 and 0.1 mg readability models feature a draft shied (which can not be removed). The 0.001g readability models feature a removable, glass shield. All units are supplied with switching AC adapter, weighing pan, weighing pan supports, user manual, and quick start guide. Includes five-year warranty. Accessories not included. Drop shipment only. Customer information required for calibration certificate.
Precaución: Fragile and handle with care.