VWR® T-Series (NTEP) Balances, with Calibration Certificate
Supplier: VWR, part of Avantor
VWR®
VWR-160TC.N/C VWR-6100TC.N/C VWR-220TC.N/C VWR-205TC.N/C VWR-310TC.N/C VWR-8100TC.N/C VWR-4500TC.N/C VWR-105TC.N/C
76458-502EA 8793.63 USD
76458-502 76458-514 76458-512 76458-506 76458-504 76458-516 76458-508 76458-510
VWR® T-Series (NTEP) Balances, with Calibration Certificate
Balances and Scales
The VWR® T-Series balances feature state-of-the-art technology to provide the highest accuracy measurements.
  • Large, 5" color touch screen
  • Touchless operation via IR proximity sensors
  • Internal and external calibration
  • Seven databases
These balances provide unsurpassed performance ideal for any laboratory condition. These user-friendly balances offer a variety of standard features, such as GLP procedures, 17 weighing units, adjustable ambient condition settings, 11 application modes, and quick stabilization time. These balances are the answer for organizations looking for a user-friendly balance that provides the highest quality results.
Certificaciones: NTEP certified.