VWR® T-Series Balances with Calibration Certificate
Supplier: VWR, part of Avantor
VWR®
VWR-363TC/C VWR-520TC/C VWR-105TC/C VWR-3003TC/C VWR-2102TC/C VWR-1003TC/C VWR-4502TC/C VWR-205TC/C VWR-160TC/C VWR-10102TC/C VWR-603TC/C VWR-8102TC/C VWR-310TC/C VWR-225TC/C VWR-6002TC/C VWR-220TC/C
76378-460EA 2268.04 USD
76378-460 76378-472 76378-470 76458-490 76378-468 76458-494 76378-466 76458-492 76458-498 76378-464 76458-488 76458-496 76378-462 76458-486 76458-500 76378-438
VWR® T-Series Balances with Calibration Certificate
Balances and Scales
The VWR® T-Series balances with calibration certificate are the answer for an organizations regulatory and compliance needs.
  • Complete calibration data reported
  • 2×RS232, USB (A and B), Ethernet and Wi-Fi communication interfaces
  • Internal calibration
  • Large, 5" color touch screen
  • Touchless operation via IR proximity sensors
  • Tool-free assembly and disassembly of the three-door draft shield
  • 5-year warranty
The VWR® T-Series balances with calibration certificate include an ISO 17025:2017 certificate of calibration showing the full testing data. These balances provide accuracy and precision for any laboratory environment. Featuring internal calibration, IR proximity sensors for hands free operation, a multitude of communication options, and built in data protections, and provides advanced features for any application.
The large, color touch screen allows for easy viewing of results and navigation through the balances menus. Data safety is provided via the internal memory allows for recording up to 500,000 weighings. The analytical models feature a spacious 3-door draft shield that allows for tool-free assembly and disassembly of the weighing chamber. With its 5-year warranty, this balance will provide reliable results for years to come.
The balances include a certificate of calibration showing the full testing data from an ISO/IEC 17025:2017 accredited calibration and testing laboratory utilizing NIST-traceable weights.
Certificaciones: ISO/IEC 17025:2017 accredited calibration and testing laboratory issues calibration certificate, CE marked.
Información para pedidos: All units supplied with switching AC adapter, weighing pan, weighing pan supports, user manual, and quick start guide. Corresponding NTEP certified legal for trade models are also available, please see vwr.com for details. Includes five-year warranty. Accessories not included.