Avantor® Hichrom HI-SE54 GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI0532010105 HI05-10-010-10 HI05-10-010-15 HI05-32-025-50 HI05-53-265-10 HI05-20-010-60 HI05-53-100-75 HI05-05-005-2 HI0532025105 HI05-32-300-50 HI05-25-025-50 HI05-18-040-10 HI05-25-010-50 HI05-20-020-60 HI05-10-020-10 HI05-53-100-10 HI05-32-400-30 HI05-20-020-50 HI05-10-040-5 HI05-32-010-60 HI05-32-025-60 HI05-32-050-30 HI05-32-150-30 HI0525025105 HI05-53-300-75 HI05-18-018-10 HI05-20-100-10 HI05-05-005-5 HI05-25-100-15 HI05-53-100-30 HI05-18-018-20 HI05-53-025-30 HI05-20-020-20 HI05-18-018-40 HI05-15-015-15 HI05-18-020-20 HI05050052.5 HI05-53-700-30 HI05-25-050-15 HI0532150105 HI05-32-150-10 HI05-53-025-50 HI05-32-025-15 HI05-53-010-30 HI053210030G5 HI0532050105 HI05-32-100-60 HI05-25-050-60 HI05-53-150-75 HI0525100105 HI05-32-050-50 HI05-20-010-50 HI05-25-025-60 HI05-32-100-10 HI05-18-020-60 HI05-32-150-15 HI05-32-100-30 HI05-25-100-60 HI05-18-020-50 HI05-53-700-15 HI05-32-100-50 HI05-18-040-50 HI05-18-020-40 HI05-53-300-60 HI05-20-040-40 HI05-18-040-40 HI05-53-300-15 HI05-18-018-60 HI05-53-100-15 HI053201010G2 HI05-53-150-30 HI0532300105 HI05-32-010-50 HI0532100105 HI05-32-050-60 HI05-32-150-60 HI05-53-500-10 HI05-10-010-5 HI05-53-050-10 HI05-25-050-50 HI05-32-500-60 HI05-25-010-15 HI05-32-500-50 HI05-32-300-60 HI05-53-010-10 HI05-25-050-30 HI05-53-150-60 HI05-20-100-40 HI05-15-015-20 HI055305030G10 HI05-53-150-50 HI05-53-150-15 HI05320108G2 HI05-53-500-30 HI05-53-300-50 HI05-53-100-60 HI05-53-050-60 HI05-53-300-30 HI05-25-010-30 HI05-18-018-50 HI0532500105 HI05-18-020-10 HI05-20-010-20 HI052502515G5 HI05-32-010-15 HI05-10-040-15 HI05-32-300-15 HI05-20-010-40 HI05-53-050-15 HI05-25-010-60 HI05-25-100-50 HI05-32-025-10 HI05-32-300-30 HI05-18-040-20 HI05-53-025-15 HI05-10-020-15 HI05-32-025-30 HI05-15-015-5 HI05-25-100-30 HI05-20-020-10 HI05-15-015-30 HI05-53-010-15 HI05-20-040-60 HI05-32-100-15 HI05-53-500-75 HI05-10-040-10 HI05-20-100-20 HI05-32-050-15 HI05-53-500-60 HI05-53-150-10 HI05-53-100-50 HI05-32-010-10 HI05-25-025-15 HI05-25-100-10 HI05-15-015-10 HI05-20-100-60 HI052502560G5 HI05-53-050-50 HI05-32-500-30 HI05-53-500-15 HI05-10-020-5 HI05-32-300-10 HI05-20-020-40 HI05-20-010-10 HI05-25-025-10 HI05-53-300-10 HI05-20-040-20 HI05-18-040-60 HI052510030G5 HI052502530G5 HI05-32-150-50 HI05-20-040-50 HI05-25-025-30 HI05-25-050-10 HI05-20-040-10 HI0525050105 HI05-32-010-30 HI05-32-050-10 HI05-53-500-50 HI05-53-025-60 HI05-53-050-30 HI05-20-100-50 HI05-32-500-10 HI05-32-500-15 HI055310030G5
76435-588EA 2584.59 USD
76435-588 76430-920 76430-922 76435-606 76435-720 76435-510 76435-706 76430-914 76435-600 76435-654 76435-558 76435-492 76435-546 76435-520 76430-926 76435-696 76435-658 76435-518 76430-936 76435-596 76435-608 76435-616 76435-640 76435-552 76435-732 76435-472 76435-532 76430-918 76435-578 76435-700 76435-474 76435-680 76435-514 76435-476 76430-940 76435-484 76430-916 76435-748 76435-566 76435-636 76435-634 76435-682 76435-602 76435-676 76431-416 76435-612 76435-632 76435-572 76435-718 76435-576 76435-618 76435-508 76435-560 76435-622 76435-490 76435-638 76435-628 76435-584 76435-488 76435-746 76435-630 76435-498 76435-486 76435-730 76435-526 76435-496 76435-724 76435-480 76435-698 76431-412 76435-712 76435-648 76435-594 76435-624 76435-620 76435-644 76435-734 76430-924 76435-686 76435-570 76435-670 76435-542 76435-668 76435-656 76435-672 76435-568 76435-716 76435-536 76430-942 76431-418 76435-714 76435-710 76431-414 76435-738 76435-728 76435-704 76435-694 76435-726 76435-544 76435-478 76435-662 76435-482 76435-504 76431-404 76435-590 76430-934 76435-650 76435-506 76435-688 76435-548 76435-582 76435-598 76435-652 76435-494 76435-678 76430-928 76435-604 76430-946 76435-580 76435-512 76430-944 76435-674 76435-530 76435-626 76435-744 76430-932 76435-534 76435-614 76435-742 76435-708 76435-702 76435-586 76435-554 76435-574 76430-938 76435-540 76431-408 76435-692 76435-666 76435-736 76430-930 76435-646 76435-516 76435-502 76435-550 76435-722 76435-524 76435-500 76431-410 76431-406 76435-642 76435-528 76435-556 76435-562 76435-522 76435-564 76435-592 76435-610 76435-740 76435-684 76435-690 76435-538 76435-660 76435-664 76431-420
Avantor® Hichrom HI-SE54 GC Columns
Chromatography Columns
General purpose column for solvent impurities, PCBs, hydrocarbons, essential oils, semivolatiles, allergens, pesticides, and so on.
Max temperature limits may change depending on the film thickness.