Avantor® Hichrom HI-SE52 GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI042502530G5 HI04-20-100-20 HI04-18-018-10 HI04-32-010-50 HI04-32-300-60 HI04-15-015-15 HI04-32-025-50 HI04-25-050-10 HI04-15-015-10 HI04-53-100-30 HI04320108G2 HI04-53-025-15 HI0432050105 HI04-18-020-50 HI04-32-010-10 HI04-32-300-30 HI04-53-050-15 HI04-18-018-40 HI04-53-100-60 HI04-15-015-20 HI04-10-040-15 HI04-15-015-5 HI04-32-050-10 HI04-32-500-30 HI04-53-300-50 HI04-18-018-60 HI04-15-015-30 HI04-32-050-30 HI04-32-025-15 HI0432500105 HI04-20-100-50 HI04-53-700-30 HI04-53-300-75 HI04-25-010-60 HI04-32-010-15 HI04-53-500-50 HI04-32-100-30 HI04-20-020-60 HI04-25-050-60 HI04-25-100-50 HI04-20-040-10 HI04-32-300-10 HI04-32-100-60 HI04-25-025-10 HI04-32-100-50 HI04-20-020-10 HI04-32-010-30 HI042502560G5 HI04-53-300-10 HI04-18-020-10 HI04-18-040-20 HI04-32-050-50 HI04-20-010-50 HI04-25-050-30 HI04-53-050-30 HI04-32-500-15 HI04-32-100-15 HI0425050105 HI04-25-025-15 HI04-18-020-60 HI04-32-500-60 HI04-32-400-30 HI04-18-020-40 HI04-18-040-60 HI04-32-500-50 HI04-20-010-60 HI04-10-010-5 HI04-20-020-40 HI04-25-050-15 HI04-32-025-30 HI04-25-025-60 HI04-53-300-60 HI04-53-500-60 HI04-32-150-30 HI04-10-040-5 HI042502515G5 HI04-25-025-30 HI04-32-010-60 HI042510030G5 HI04-25-025-50 HI04-10-020-5 HI04-20-100-40 HI04-25-050-50 HI04-25-010-50 HI04-53-150-30 HI04-25-100-30 HI04-10-020-10 HI04-53-025-50 HI04-05-005-2 HI04-20-010-20 HI04-53-265-10 HI04-32-300-15 HI04-20-040-60 HI043210030G5 HI04-32-150-50 HI04-53-025-60 HI04-53-050-10 HI04-18-018-20 HI04-53-010-30 HI04-53-100-75 HI04-10-020-15 HI04-53-150-10 HI04-53-050-60 HI04-25-010-30 HI04-32-050-60 HI04-53-300-15 HI0432010105 HI0425025105 HI04-53-025-30 HI04-18-040-10 HI04-53-150-75 HI04-53-150-60 HI04-53-150-15 HI04-05-005-5 HI04-32-100-10 HI04-32-150-15 HI04-20-040-20 HI04-32-150-10 HI04-10-010-15 HI04-53-500-10 HI04-20-100-60 HI04-25-100-15 HI04-53-050-50 HI04-32-300-50 HI04-20-100-10 HI0432300105 HI04-32-025-60 HI04050052.5 HI04-53-010-15 HI0425100105 HI04-25-100-10 HI04-32-050-15 HI04-53-100-50 HI04-53-300-30 HI04-53-500-30 HI04-18-020-20 HI04-32-150-60 HI045305030G10 HI0432100105 HI04-53-500-15 HI04-20-010-40 HI04-25-100-60 HI04-20-040-40 HI043201010G2 HI04-18-040-50 HI04-10-010-10 HI04-20-020-20 HI04-32-025-10 HI0432025105 HI045310030G5 HI04-53-700-15 HI04-53-100-10 HI04-53-010-10 HI04-10-040-10 HI04-20-010-10 HI04-20-020-50 HI04-20-040-50 HI04-53-500-75 HI04-25-010-15 HI04-32-500-10 HI04-18-018-50 HI04-53-150-50 HI04-18-040-40 HI04-53-100-15 HI0432150105
76431-388EA 1044.12 USD
76431-388 76435-256 76435-194 76435-316 76435-378 76430-906 76435-328 76435-284 76430-904 76435-422 76431-396 76435-400 76435-334 76435-210 76435-308 76435-374 76435-410 76435-198 76435-426 76430-908 76430-900 76430-912 76435-332 76435-388 76435-450 76435-202 76430-910 76435-338 76435-324 76435-384 76435-260 76435-470 76435-454 76435-270 76435-312 76435-462 76435-350 76435-242 76435-294 76435-304 76435-244 76435-368 76435-354 76435-272 76435-352 76435-234 76435-314 76431-390 76435-444 76435-204 76435-216 76435-340 76435-230 76435-290 76435-412 76435-386 76435-348 76435-286 76435-276 76435-212 76435-392 76435-380 76435-208 76435-222 76435-390 76435-232 76430-890 76435-238 76435-288 76435-326 76435-282 76435-452 76435-464 76435-362 76430-902 76431-386 76435-278 76435-318 76431-392 76435-280 76430-896 76435-258 76435-292 76435-268 76435-434 76435-302 76430-892 76435-404 76430-880 76435-226 76435-442 76435-372 76435-252 76431-398 76435-364 76435-406 76435-408 76435-196 76435-398 76435-428 76430-894 76435-430 76435-416 76435-266 76435-342 76435-446 76435-310 76435-274 76435-402 76435-214 76435-440 76435-438 76435-432 76430-884 76435-344 76435-360 76435-246 76435-356 76430-888 76435-456 76435-262 76435-300 76435-414 76435-376 76435-254 76435-370 76435-330 76430-882 76435-396 76435-298 76435-296 76435-336 76435-424 76435-448 76435-460 76435-206 76435-366 76431-400 76435-346 76435-458 76435-228 76435-306 76435-248 76431-394 76435-220 76430-886 76435-236 76435-320 76435-322 76431-402 76435-468 76435-418 76435-394 76430-898 76435-224 76435-240 76435-250 76435-466 76435-264 76435-382 76435-200 76435-436 76435-218 76435-420 76435-358
Avantor® Hichrom HI-SE52 GC Columns
Chromatography Columns
General purpose column for solvent impurities, PCBs, hydrocarbons, essential oils, semivolatiles, triglycerides, pesticides, poly-waxes, and so on.
Maximum temperature limits may change depending on the film thickness.