Avantor® Hichrom HI-1701 GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI0625100105 HI06-53-300-60 HI06-25-100-30 HI06-10-010-5 HI06-18-020-10 HI06-32-010-30 HI06-53-050-30 HI06-20-018-10 HI06-15-015-30 HI06-32-050-10 HI06-53-300-30 HI06-53-025-30 HI06-20-020-10 HI06-25-050-30 HI06-20-040-20 HI06-10-020-10 HI06-25-010-60 HI06-25-050-15 HI06-32-050-15 HI06-32-025-30 HI062502560G5 HI06-25-100-15 HI0632050105 HI06-53-050-15 HI06-18-020-40 HI06-32-150-10 HI06-10-010-10 HI06-32-300-60 HI06-53-300-75 HI06-25-010-50 HI06-53-300-15 HI06-53-025-15 HI06-18-040-40 HI06-20-020-50 HI06-25-025-10 HI06-53-100-30 HI06-32-025-10 HI0632025105 HI06-20-040-40 HI06-18-040-60 HI06-32-010-10 HI06-20-020-40 HI06-25-010-15 HI06-32-300-10 HI06-32-100-10 HI06-20-040-60 HI06-10-040-15 HI06-32-010-50 HI062502530G5 HI06-20-018-40 HI06-53-025-50 HI06-32-100-15 HI06-18-018-60 HI06-53-150-15 HI06-53-300-50 HI06-53-100-50 HI0632300105 HI06-25-025-60 HI0625050105 HI06-32-300-30 HI06-18-018-10 HI06-15-015-10 HI06-53-150-60 HI06-53-150-75 HI06-32-100-50 HI06-18-020-50 HI06-32-150-15 HI06-20-040-10 HI06-53-050-50 HI06-32-010-15 HI06-15-015-20 HI06-32-050-30 HI06-10-020-5 HI0632150105 HI06-32-025-50 HI06-18-018-20 HI06-32-050-50 HI065310030G5 HI06-25-025-30 HI06-05-005-5 HI06-32-300-15 HI06-10-040-10 HI06-20-020-20 HI06-32-025-60 HI06-25-050-60 HI06-32-150-50 HI0632010105 HI06-20-018-50 HI06-25-010-30 HI06-15-015-15 HI06-32-100-30 HI06-53-300-10 HI062502515G5 HI06-32-100-60 HI06-05-005-2 HI06-32-150-30 HI06-53-150-10 HI06-25-025-50 HI06-10-020-15 HI06-10-010-15 HI0625025105 HI06-53-100-15 HI06-25-100-60 HI06-20-020-60 HI06-53-150-30 HI06-32-025-15 HI06-15-015-5 HI0632100105 HI06-18-040-20 HI06-32-050-60 HI06-20-018-20 HI06-53-050-10 HI06-53-010-15 HI06-18-018-50 HI06-53-025-60 HI06-32-300-50 HI06-53-150-50 HI06-18-020-60 HI06-18-040-10 HI06-10-040-5 HI06-25-100-50 HI06-53-050-60 HI06-20-040-50 HI06-25-050-50 HI06-53-100-75 HI06-53-010-30 HI06-32-150-60 HI06-18-020-20 HI06-25-100-10 HI06-53-100-60 HI06-53-100-10 HI06-18-040-50 HI06-25-025-15 HI06-32-010-60 HI06-25-050-10 HI06-53-010-10 HI06050052.5 HI06-20-018-60 HI06-18-018-40
76434-770EA 2525.82 USD
76434-770 76434-908 76434-774 76430-250 76434-686 76434-786 76434-870 76434-706 76430-270 76434-804 76434-904 76434-860 76434-716 76434-762 76434-728 76430-252 76434-742 76434-760 76434-808 76434-798 76431-094 76434-772 76434-806 76434-868 76434-690 76434-828 76430-246 76434-850 76434-910 76434-740 76434-902 76434-858 76434-700 76434-722 76434-744 76434-880 76434-792 76434-794 76434-730 76434-704 76434-780 76434-720 76434-736 76434-840 76434-816 76434-734 76430-260 76434-788 76431-092 76434-710 76434-862 76434-820 76434-684 76434-890 76434-906 76434-882 76434-842 76434-754 76434-758 76434-846 76434-676 76430-264 76434-896 76434-898 76434-824 76434-692 76434-832 76434-726 76434-872 76434-784 76430-268 76434-810 76430-256 76434-830 76434-800 76434-678 76434-812 76431-096 76434-750 76430-244 76434-844 76430-258 76434-718 76434-802 76434-766 76434-836 76434-782 76434-712 76434-738 76430-266 76434-822 76434-900 76431-090 76434-826 76430-240 76434-834 76434-888 76434-752 76430-254 76430-248 76434-746 76434-878 76434-778 76434-724 76434-892 76434-796 76430-272 76434-818 76434-698 76434-814 76434-708 76434-866 76434-854 76434-682 76434-864 76434-848 76434-894 76434-694 76434-696 76430-262 76434-776 76434-874 76434-732 76434-764 76434-886 76434-856 76434-838 76434-688 76434-768 76434-884 76434-876 76434-702 76434-748 76434-790 76434-756 76434-852 76430-242 76434-714 76434-680
Avantor® Hichrom HI-1701 GC Columns
Chromatography Columns
General purpose low-to-mid-polarity phase for a wide range of applications, including Aroclors, pesticides, herbicides and more.
  • Low bleed
  • Long lifetime
Maximum temperature limits may change depending on the film thickness.