Avantor® Hichrom HI-17 GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI07-10-010-5 HI07-32-025-30 HI07-53-150-50 HI07-53-025-50 HI07-25-025-50 HI07-53-050-50 HI0732100105 HI07-10-020-5 HI07-20-020-50 HI07-32-050-10 HI07-25-010-15 HI07-53-150-75 HI07-20-020-40 HI07-53-300-15 HI07-32-150-15 HI07-18-020-10 HI07-25-100-30 HI07-53-100-75 HI07-20-010-10 HI072502530G5 HI07-18-040-50 HI07-25-050-10 HI07-25-010-50 HI07-32-150-50 HI07-53-150-15 HI07-20-010-40 HI07-53-300-10 HI0732300105 HI0725025105 HI07-18-020-20 HI07-32-010-10 HI07-32-010-60 HI07-53-150-30 HI07-53-010-10 HI07-32-050-15 HI07-53-025-30 HI07-53-010-30 HI07-32-300-30 HI07-25-050-50 HI07-53-100-30 HI07-53-265-30 HI0732150105 HI07-15-015-15 HI07-20-010-20 HI07-25-025-10 HI07-20-040-50 HI07-15-015-10 HI07-10-010-15 HI07-25-025-15 HI07-53-010-15 HI07-20-010-50 HI07-10-040-15 HI07-20-040-10 HI07-15-015-20 HI07-53-100-60 HI07-15-015-5 HI07-20-040-60 HI07-32-100-10 HI07-32-150-30 HI07-53-050-30 HI07-20-100-10 HI0725100105 HI07-18-020-50 HI07-32-025-60 HI07-32-050-60 HI07-32-050-50 HI07-18-020-40 HI07-20-100-40 HI07-25-100-10 HI0725050105 HI07-20-020-60 HI07050052.5 HI07-18-018-10 HI07-25-050-60 HI07-53-025-60 HI07-25-100-15 HI07-32-100-30 HI07-32-100-60 HI07-10-040-5 HI07-53-050-10 HI07-32-025-50 HI07-10-040-10 HI07-53-050-15 HI07-32-010-15 HI07-25-010-30 HI07-05-005-5 HI07-32-300-60 HI07-25-050-30 HI07-53-100-50 HI0732025105 HI07-10-020-15 HI07-53-300-30 HI07-25-025-30 HI07-20-100-20 HI07-20-010-60 HI07-32-100-15 HI072502560G5 HI07-32-100-50 HI07-32-050-30 HI07-53-025-15 HI07-53-300-50 HI07-20-040-40 HI07-53-150-10 HI07-32-010-50 HI07-53-265-10 HI07-25-025-60 HI07-20-100-60 HI07-32-025-10 HI07-15-015-30 HI07-05-005-2 HI07-25-100-50 HI07-20-020-10 HI07-18-040-20 HI07-32-150-10 HI07-18-018-50 HI075310030G5 HI07-32-300-10 HI07-25-100-60 HI07-10-010-10 HI0732050105 HI07-25-010-60 HI072502515G5 HI07-53-300-75 HI07-20-020-20 HI07-32-010-30 HI07-25-050-15 HI07-10-020-10 HI07-53-100-10 HI07-18-020-60 HI07-32-300-15 HI07-18-018-20 HI07-32-025-15 HI07-18-040-60 HI07-18-040-10 HI07-20-040-20 HI07-32-300-50 HI0732010105 HI07-18-018-40 HI07-18-018-60 HI07-53-100-15 HI07-53-150-60 HI07-18-040-40 HI07-53-300-60 HI07-32-150-60 HI07-53-050-60
76430-206EA 578.61 USD
76430-206 76434-580 76434-460 76430-208 76434-582 76434-462 76434-584 76434-464 76434-586 76434-620 76434-466 76434-500 76434-588 76434-622 76434-468 76434-458 76434-612 76434-614 76434-616 76434-618 76434-570 76434-450 76434-572 76434-452 76434-574 76434-454 76434-576 76434-610 76434-456 76434-578 76434-568 76434-602 76434-448 76434-604 76434-606 76434-608 76430-224 76430-226 76430-228 76434-560 76434-440 76434-562 76434-442 76434-564 76434-444 76430-220 76434-566 76430-222 76434-600 76434-446 76434-436 76434-558 76434-438 76430-212 76430-214 76430-216 76430-218 76434-670 76434-550 76434-430 76434-672 76434-552 76434-674 76434-432 76434-554 76430-210 76434-434 76434-556 76431-088 76434-546 76434-668 76431-086 76434-548 76434-428 76431-084 76431-082 76434-660 76434-540 76434-662 76434-542 76434-664 76434-544 76434-666 76434-656 76434-536 76434-658 76434-538 76430-234 76430-236 76430-238 76434-490 76434-492 76434-494 76434-496 76434-650 76434-530 76434-652 76430-230 76434-498 76434-532 76434-654 76430-232 76434-534 76434-524 76434-646 76434-526 76434-648 76434-528 76434-480 76434-482 76434-484 76434-640 76434-486 76434-520 76434-488 76434-642 76434-522 76434-644 76434-634 76434-514 76434-636 76434-516 76434-638 76434-518 76434-590 76434-470 76434-592 76434-472 76434-594 76434-474 76434-596 76434-630 76434-476 76434-510 76434-598 76434-632 76434-478 76434-512 76434-502 76434-624 76434-504 76434-626 76434-506 76434-628 76434-508
Avantor® Hichrom HI-17 GC Columns
Chromatography Columns
General purpose polar phase for a wide range of applications, ideal as confirmation analysis column. Ideal in a GCxGC configuration as polar dimension column. HI-17 is a Crossbonded phase and assure a long lifetime even under high temperature limits.
  • Phase composition: crossbonded 50% Phenyl, 50% Methyl Polysiloxane
  • Long lifetime
Max temperature limits may change depending on the film thickness.