Avantor® Hichrom HI-17 MS GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI22-18-040-60 HI22-18-020-10 HI22-53-150-15 HI22-53-150-50 HI22-53-150-30 HI22-10-010-15 HI22-53-050-15 HI22050052.5 HI22-53-050-60 HI22-20-033-12 HI22-20-100-10 HI22-25-050-30 HI22-32-025-30 HI22-32-010-50 HI22-20-020-60 HI22-32-050-30 HI22-32-025-50 HI22-53-025-60 HI22-53-100-60 HI22-53-025-30 HI22-18-040-20 HI22-32-010-30 HI22-20-033-25 HI222502515G5 HI22-20-033-50 HI22-15-015-30 HI22-25-100-60 HI222505030G10 HI22-18-036-20 HI22-18-036-40 HI22-18-018-20 HI22-20-100-60 HI22-20-100-40 HI22-53-010-30 HI2232150105 HI22-25-025-30 HI22-10-020-15 HI22-25-100-30 HI22-53-025-50 HI22-05-005-2 HI22-20-010-60 HI22-53-050-30 HI22-25-025-10 HI22-15-015-5 HI22-25-025-60 HI22-20-100-20 HI223210030G5 HI22-32-100-30 HI22-32-100-15 HI22-53-100-15 HI22-53-010-15 HI222502560G10 HI22-25-025-15 HI22-25-010-30 HI222502560G5 HI22-18-018-60 HI22-10-040-5 HI2225025105 HI22-32-150-50 HI22-18-018-10 HI22-53-050-10 HI22-25-050-50 HI223202530G5 HI22-53-100-10 HI22-20-040-50 HI22-32-025-15 HI22-18-040-50 HI222505030G5 HI22-18-020-40 HI22-20-100-50 HI22-20-010-50 HI22-20-040-60 HI22-32-025-10 HI22-53-025-15 HI22-20-020-20 HI2225050105 HI222505015G10 HI22-10-010-10 HI22-53-100-75 HI22-25-010-50 HI22-18-040-40 HI2232050105 HI225310030G5 HI22-10-010-5 HI22-32-100-10 HI22-32-025-60 HI2225100105 HI22-32-100-60 HI22-25-025-50 HI22-20-033-60 HI222502530G5 HI22-32-010-10 HI22-32-050-10 HI22-18-020-50 HI22-32-010-15 HI22-32-150-30 HI22-53-150-75 HI22-18-018-50 HI22-20-010-10 HI22-25-010-15 HI22-32-150-10 HI22-32-050-60 HI22-20-040-10 HI22-32-050-50 HI22-32-100-50 HI22-53-150-10 HI22-10-040-10 HI22-53-100-50 HI22-18-036-60 HI22-20-020-10 HI22-32-150-60 HI22-25-050-10 HI22-25-050-60 HI22-15-015-15 HI22-32-050-15 HI22-32-010-60 HI22-15-015-10 HI22-25-100-50 HI2232100105 HI22-20-033-30 HI22-53-050-50 HI2232025105 HI22-20-010-40 HI22-18-040-10 HI22-05-005-5 HI22-25-100-10 HI22-25-050-15 HI222501030G5 HI22-20-020-50 HI222502530G10 HI22-25-010-60 HI22-18-020-20 HI22-53-150-60 HI22-20-020-40 HI22-18-020-60 HI22-20-040-20 HI22-20-040-40 HI22-18-018-40 HI22-53-010-10 HI22-32-150-15 HI22-53-100-30 HI2232010105 HI22-10-020-10 HI22-10-020-5 HI22-15-015-20 HI22-10-040-15 HI22-25-100-15 HI22-20-010-20
76432-294EA 1688.79 USD
76432-294 76432-174 76432-296 76432-176 76432-290 76432-170 76432-292 76432-172 76432-214 76432-216 76432-210 76432-298 76432-178 76432-212 76431-098 76432-208 76432-184 76433-888 76432-186 76432-220 76432-180 76432-182 76432-104 76432-226 76432-106 76432-228 76432-100 76432-188 76432-222 76432-102 76432-224 76432-218 76433-890 76433-894 76433-892 76433-898 76433-896 76432-272 76433-912 76432-152 76432-274 76433-910 76432-154 76433-916 76432-270 76433-914 76432-150 76433-918 76432-276 76432-156 76432-278 76432-158 76431-110 76432-306 76432-308 76431-118 76431-116 76433-886 76431-114 76431-112 76432-162 76432-284 76433-902 76432-164 76433-900 76432-286 76432-280 76433-906 76432-160 76433-904 76432-282 76432-204 76433-908 76432-206 76432-166 76432-200 76432-288 76432-168 76432-202 76431-120 76432-250 76432-096 76432-130 76432-252 76432-098 76432-132 76432-092 76432-094 76432-258 76432-138 76432-254 76432-134 76432-256 76432-136 76432-090 76432-140 76432-262 76432-142 76432-264 76432-260 76432-302 76432-148 76432-304 76432-144 76432-266 76432-300 76432-146 76432-268 76431-100 76431-108 76431-106 76431-104 76431-102 76432-074 76432-196 76432-230 76432-076 76432-198 76432-110 76432-192 76432-072 76432-194 76432-236 76432-116 76432-238 76432-118 76432-232 76432-078 76432-112 76432-234 76432-114 76432-108 76432-190 76432-086 76432-240 76432-120 76432-242 76432-088 76432-082 76432-084 76432-126 76432-248 76432-128 76432-122 76432-244 76432-124 76432-246 76432-080
Avantor® Hichrom HI-17 MS GC Columns
Chromatography Columns
Really versatile general purpose polar phase for a wide range of applications. Ideal in confirmation analysis. HI-17 MS is an MS grade low-bleeding crossbonded column and assure a long lifetime even under high temperature limits.
  • Phase composition: Crossbonded Silphenylene - Methyl Polysiloxane (17MS column selectivity) - MS grade
  • Long lifetime
Max temperature limits may change depending on the film thickness.